1,8-Diiodooctane ≥97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24772-63-2
:
Formula:C₈H₁₆I₂
MDL Number:MFCD00001106
:366.02 g/mol
Boiling Pt:167…169 °C (6 mmHg)
Melting Pt:16…21 °C
Density:1,840
Flash Pt:>110 °C (230 °F)
:Stabilised with copper
EINECS:246-456-6
:GHS07
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Organic Photovoltaics
Product Literature
Frequently Bought Together