Magnesium chloride hexahydrate 99.0-101.0% (by complexometric analysis), EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
: Merck
: Magnesium dichloride
:7791-18-6
:
:MgCl₂·6H₂O
:MFCD00149781
:1412
:117
:1.57 g/cm³ (25 °C)
:203.3 g/mol
:13,05684
:M-Clarity™ quality level = MQ300
:Ambient
:232-094-6
- Environmentally Preferable:
- Assay (complexometric):99.0 - 101.0 %
- Identity:conforms
- Appearance of solution:passes test
- Insoluble matter:≤ 0.005 %
- pH-value (5 %; water):5.0 - 6.5
- Acidity or alkalinity:passes test
- Bromide (Br):≤ 0.05 %
- Nitrate (NO₃):≤ 0.001 %
- Phosphate (PO₄):≤ 0.0005 %
- Sulfate (SO₄):≤ 0.002 %
- Total nitrogen (N):≤ 0.0002 %
- Heavy metals (as Pb):≤ 0.0005 %
- Al (Aluminium):≤ 0.0002 %
- As (Arsenic):≤ 0.0002 %
- Ba (Barium):≤ 0.002 %
- Ca (Calcium):≤ 0.003 %
- Cu (Copper):≤ 0.0005 %
- Fe (Iron):≤ 0.0005 %
- K (Potassium):≤ 0.001 %
- Mn (Manganese):≤ 0.0005 %
- Na (Sodium):≤ 0.001 %
- NH₄ (Ammonium):≤ 0.002 %
- Pb (Lead):≤ 0.0005 %
- Sr (Strontium):≤ 0.005 %
- Water:51.0 - 55.0 %
Frequently Bought Together