
Sodium hydrogen sulphate monohydrate ≥99.0% (by titrimetric analysis), Puriss. p.a.
Catalog # 71657-500G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay:99.0 - 101.0 %
- Silver (Ag):Max. 5 ppm
- Aluminium (Al):Max. 5 ppm
- Barium (Ba):Max. 5 ppm
- Bismuth (Bi):Max. 5 ppm
- Calcium (Ca):Max. 10 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 20 ppm
- Lithium (Li):Max. 5 ppm
- Magnesium (Mg):Max. 5 ppm
- Manganese (Mn):Max. 5 ppm
- Molybdenum (Mo):Max. 5 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Strontium (Sr):Max. 5 ppm
- Thallium (Tl):Max. 5 ppm
- Zinc (Zn):Max. 5 ppm
- Chloride (Cl):Max. 10 ppm
- Phosphate (PO₄):Max. 5 ppm
- Total N:Max. 10 ppm
Specification Test Results
About this item
:
:138.08 g/mol
Merck Index:13,08658
Boiling Pt:Decomposes on heating
Boiling Pt: Dec. (on heating)
Melting Pt:58 °C
Density:2,12 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-665-7
ADR:8,III
:GHS05
CAS Number:10034-88-5
Formula:NaHSO₄·H₂O
MDL Number:MFCD00149210