
Sodium hydrogen sulphate monohydrate 97%, crystals, extra pure reagent
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10034-88-5
:
Formula:NaHSO₄·H₂O
MDL Number:MFCD00149210
:138.08 g/mol
Merck Index:13,08658
Melting Pt:58 °C
Density:2,1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-665-7
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White Adhering crystals
- Titration with NaOH:≥96.0 %
- Chloride (Cl):≤20 ppm
- Iron (Fe):≤50 ppm
- Heavy metals (as Pb):≤50 ppm
Specification Test Results
Frequently Bought Together