Sodium sulphate decahydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7727-73-3
:
Formula:Na₂SO₄·10H₂O
MDL Number:MFCD00149189
:322.2 g/mol
Merck Index:14,8680
Boiling Pt:1429 °C (1013 hPa)
Melting Pt:∼32 °C
Density:1,460
Storage Temperature:Ambient
EINECS:231-820-9
Related Information
Frequently Bought Together