Sodium perchlorate monohydrate ≥85.0-90.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7791-07-3
:
Formula:NaClO₄·H₂O
MDL Number:MFCD00149164
:140.46 g/mol
Merck Index:14,8653
Melting Pt:130 °C (dec.)
Density:2,02
Storage Temperature:Ambient
EINECS:231-511-9
UN:1502
ADR:5.1,II
:GHS03
:GHS07
Related Information
Frequently Bought Together