
Sodium carbonate monohydrate ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5968-11-6
:
Formula:Na₂CO₃·1H₂O
MDL Number:MFCD00149177
:124.00 g/mol
Merck Index:14,8596
Melting Pt:100 °C (-H₂O)
Density:2,250
EINECS:207-838-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together