Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
(R)-(-)-Epichlorhydrin 99%

(R)-(-)-Epichlorhydrin 99%

Toxic
Health
Corrosive
Flammable
(R)-(-)-Epichlorhydrin 99%
Certificates
(R)-(-)-Epichlorhydrin 99%

3 Options Available Below

Product Details & Documents
Synonyms:  (R)-(-)-Glycerol epichlorhydrin, (R)-(-)-3-Chloro-1,2-epoxypropane, (R)-(-)-1-Chloro-2,3-epoxypropane
  • CAS Number:51594-55-9
  • Formula:ClCH₂CH(O)CH₂
  • MDL Number:MFCD00077759
  • Molecular Weight:92.52 g/mol
  • Boiling Pt:114 °C (760 mmHg)
  • Melting Pt:–48 °C
  • Density:1,183 g/cm³
  • Flash Pt:33 °C
  • Storage Temperature:Ambient
  • EINECS:424-280-2
  • UN:2023
  • ADR:6.1,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4360 to 1.4400 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Specific optical rotation:
    -34° to -39° (20°C, 589 nm) (c=1, MeOH)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - £91.00
    £91.00
    1 * 1 g
       £91.00

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - £188.00
    £188.00
    1 * 5 g
       £188.00

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £579.00
    £579.00
    1 * 25 g
       £579.00