
Sodium hydroxide 1 mol/l (1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur., Reag. USP, NF volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Density:1,04 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.998 - 1.002 mol/l
- Conforms to Reag. Ph.Eur.:Passes test
- Conforms to Reag. USP-NF:Passes test
- Conforms to BDH 19139:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number10L Cubitainer UN (ZPACK13260004) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
spec_VWRC31627.290 spec_VWRC31627.368 spec_VWRC31627.404
Product specification
Frequently Bought Together