Sodium dodecyl sulphate (SDS) 0.004 mol/l in aqueous solution
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
CAS Number:151-21-3
:
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
Density:1…1.1 g/cm³ (20 °C)
Molecular Weight:288.38 g/mol
Merck Index:14,08636
Storage Temperature:Ambient
EINECS:205-788-1
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together