Hydrochloric acid concentrate for dilution to 1 mol/l in aqueous solution, Titrisol®, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Molecular Weight:36.46 g/mol
ADR:8
:M-Clarity™ quality level = MQ100
EINECS:231-595-7
UN:1789
:GHS05
:GHS07
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.09 g/cm3 (20 °C)
- pH:- 1 (H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Certificates
Search by Lot NumberMerck Brighter titration Titripur®
Product Literature
msds_109970
SDS
ProductPage_109970
Product specification
Frequently Bought Together