Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
(±)-1,3-Butanediol ≥99%

(±)-1,3-Butanediol ≥99%

(±)-1,3-Butanediol ≥99%
Certificates
(±)-1,3-Butanediol ≥99%
(±)-1,3-Butanediol ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 107-88-0 Molecular Formula: C4H10O2 Formula Weight: 90.12 Flash Point: 109°(249°F) Refractive Index: 1.4400 Odour: Mild Melting Point: -50° Boiling Point: 203-204° Beilstein Reference: 1731276 Merck Reference: 14,1567 MDL No.: MFCD00004554

Synonyms:  Butane-1,3-diol, (±)-1,3-Butylene glycol, (±)-1,3-Dihydroxy butane
  • CAS Number:107-88-0
  • Formula:C₄H₁₀O₂
  • MDL Number:MFCD00004554
  • Molecular Weight:90.12 g/mol
  • Merck Index:14,1567
  • Boiling Pt:203…204 °C
  • Melting Pt:–50 °C
  • Density:1,005
  • Flash Pt:109 °C (249 °F)
  • Storage Temperature:Ambient
  • EINECS:203-529-7
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.