ContactUsLinkComponent
Anopore inorganic membranes without support ring, Whatman™
Anopore inorganic membranes without support ring, Whatman™
Certificates
undefined
Anopore inorganic membranes without support ring, Whatman™
Specifications
  • Catalog No:
  • 514-0663
  • 514-0521
  • 514-0522
  • 514-0523
  • 514-0662
  • Type:
  • Anodisc™ 47, unsupported
  • Anodisc™ 13, unsupported
  • Anodisc™ 13, unsupported
  • Anodisc™ 13, unsupported
  • Anodisc™ 47, unsupported
  • Pore size:
  • 0,20 µm
  • 0,02 µm
  • 0,10 µm
  • 0,20 µm
  • 0,02 µm
  • Thickness:
  • 60 µm
  • 60 µm
  • 60 µm
  • 60 µm
  • 60 µm
  • Max. temperature:
  • 40 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 40 °C
  • Porosity:
  • 25 - 50%
  • 25 - 50%
  • 25 - 50%
  • 25 - 50%
  • 25 - 50%
  • Refractive index:
  • 1,6
  • 1,6
  • 1,6
  • 1,6
  • 1,6