Perchloric acid 70% 70%, ULTREX® II, Ultrapure for trace metal analysis, J.T.Baker®
4806-01
: Avantor
:
- Pack type:PFA bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
- For Laboratory,Research,or Manufacturing Use:65 - 71%
- Assay (HClO₄):65 - 71 %
- Trace Impurities - Aluminum (Al):≤100 ppt
- Trace Impurities - Antimony (Sb):≤100 ppt
- Trace Impurities - Arsenic (As):≤100 ppt
- Trace Impurities - Barium (Ba):≤100 ppt
- Trace Impurities - Beryllium (Be):≤100 ppt
- Trace Impurities - Bismuth (Bi):≤10 ppt
- Trace Impurities - Cadmium (Cd):≤10 ppt
- Trace Impurities - Calcium (Ca):≤100 ppt
- Trace Impurities - Cesium (Cs):≤10 ppt
- Trace Impurities - Cobalt (Co):≤100 ppt
- Trace Impurities - Copper (Cu):≤100 ppt
- Trace Impurities - Dysprosium (Dy):≤10 ppt
- Trace Impurities - Erbium (Er):≤10 ppt
- Trace Impurities - Europium (Eu):≤10 ppt
- Trace Impurities - Gadolinium (Gd):≤10 ppt
- Trace Impurities - Gallium (Ga):≤10 ppt
- Trace Impurities - Hafnium (Hf), For Information Only:≤10 ppt
- Trace Impurities - Holmium (Ho):≤10 ppt
- Trace Impurities - Indium (In):≤10 ppt
- Trace Impurities - Iron (Fe):≤100 ppt
- Trace Impurities - Lanthanum (La):≤10 ppt
- Trace Impurities - Lead (Pb):≤10 ppt
- Trace Impurities - Lithium (Li):≤100 ppt
- Trace Impurities - Lutetium (Lu):≤10 ppt
- Trace Impurities - Magnesium (Mg):≤100 ppt
- Trace Impurities - Manganese (Mn):≤100 ppt
- Trace Impurities - Molybdenum (Mo):≤100 ppt
- Trace Impurities - Neodymium (Nd):≤10 ppt
- Trace Impurities - Nickel (Ni):≤100 ppt
- Trace Impurities - Niobium (Nb), For Information Only:≤100 ppt
- Trace Impurities - Palladium (Pd):≤10 ppt
- Trace Impurities - Platinum (Pt):≤100 ppt
- Trace Impurities - Potassium (K):≤100 ppt
- Trace Impurities - Praseodymium (Pr):≤10 ppt
- Trace Impurities - Rhodium (Rh):≤10 ppt
- Trace Impurities - Rubidium (Rb):≤10 ppt
- Trace Impurities - Samarium (Sm):≤10 ppt
- Trace Impurities - Scandium (Sc):≤100 ppt
- Trace Impurities - Silver (Ag):≤100 ppt
- Trace Impurities - Sodium (Na):≤100 ppt
- Trace Impurities - Strontium (Sr):≤100 ppt
- Trace Impurities - Tellurium (Te):≤10 ppt
- Trace Impurities - Terbium (Tb):≤10 ppt
- Trace Impurities - Thallium (Tl):≤100 ppt
- Trace Impurities - Thorium (Th):≤10 ppt
- Trace Impurities - Thulium (Tm):≤10 ppt
- Trace Impurities - Tin (Sn):≤100 ppt
- Trace Impurities - Titanium (Ti):≤100 ppt
- Trace Impurities - Tungsten (W), For Information Only:≤10 ppt
- Trace Impurities - Uranium (U):≤10 ppt
- Trace Impurities - Vanadium (V):≤100 ppt
- Trace Impurities - Ytterbium (Yb):≤10 ppt
- Trace Impurities - Yttrium (Y):≤10 ppt
- Trace Impurities - Zinc (Zn):≤100 ppt
- Trace Impurities - Zirconium (Zr):≤100 ppt
- Trace Impurities - Cerium (Ce):≤10 ppt
:
:100.46 g/mol
:231-512-4
:1873
:GHS03
:GHS05
:GHS07
:Ambient
:7601-90-3
:HClO₄
:MFCD00011325
:160…181 °C (1013 hPa)
:–112.2 °C
:1.60 g/cm³ (20 °C)
:5.1 I