

Hydrochloric acid 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1,13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,II
:36.46 g/mol
REACH:01-2119484862-27
:GHS05
:GHS07
Specification Test Results
- Assay:Min. 25.0 %
- Colouration:Max. 10 APHA
- Free chlorine:Max. 1 ppm
- Ignition residue (SO4):Max. 5 ppm
- Br (Bromide):Max. 50 ppm
- NH4 (Ammonium):Max. 1 ppm
- SO3 (Sulphite):Max. 1 ppm
- SO4 (Sulphate):Max. 2 ppm
- Al (Aluminium):Max. 0.2 ppm
- As (Arsenic):Max. 0.01 ppm
- Ba (Barium):Max. 0.05 ppm
- Be (Beryllium):Max. 0.02 ppm
- Bi (Bismuth):Max. 0.1 ppm
- Ca (Calcium):Max. 1 ppm
- Cd (Cadmium):Max. 0.01 ppm
- Co (Cobalt):Max. 0.01 ppm
- Cr (Chromium):Max. 0.02 ppm
- Cu (Copper):Max. 0.02 ppm
- Fe (Iron):Max. 0.2 ppm
- Ge (Germanium):Max. 0.05 ppm
- K (Potassium):Max. 0.1 ppm
- Li (Lithium):Max. 0.01 ppm
- Mg (Magnesium):Max. 0.2 ppm
- Mn (Manganese):Max. 0.01 ppm
- Mo (Molybdenum):Max. 0.02 ppm
- Na (Sodium):Max. 0.5 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.02 ppm
- Sr (Strontium):Max. 0.01 ppm
- Ti (Titanium):Max. 0.1 ppm
- Tl (Thallium):Max. 0.05 ppm
- V (Vanadium):Max. 0.01 ppm
- Zn (Zinc):Max. 0.1 ppm
- Zr (Zirconium):Max. 0.1 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500)
Packaging Information
Frequently Bought Together