


Formaldehyde 37% in aqueous solution stabilised, Proteomics Grade
Catalog # M134-500ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Bottle, amber
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Acidity:<0.03%
- Color (APHA):≤10
- Formaldehyde:37 – 37.5%
- Methanol:10.5 – 13%
Specification Test Results
About this item
:
:30.03 g/mol
ADR:8,III
:Stabilised with methanol 10,5 – 13%
EINECS:200-001-8
UN:1198
:GHS05
:GHS08
:GHS06
Storage Temperature:Ambient
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274