4-Methylcyclohexanol cis- and trans-mixture ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:589-91-3
:
Formula:C₇H₁₄O
MDL Number:MFCD00001449
Boiling Pt:171…173 °C
Melting Pt:–41 °C
Density:0,915
Flash Pt:70 °C (158 °F)
:114.19 g/mol
Storage Temperature:Ambient
EINECS:209-664-8
Related Information
Frequently Bought Together