Order Entry
Northern Ireland
ContactUsLinkComponent
4-Methylcyclohexanol cis- and trans-mixture 99%
Irritant
4-Methylcyclohexanol cis- and trans-mixture 99%
Certificates
undefined
Irritant
4-Methylcyclohexanol cis- and trans-mixture 99%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4570 to 1.4590 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together