

2-Methylcyclohexanol cis- and trans-mixture 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:583-59-5
:
Formula:C₇H₁₄O
MDL Number:MFCD00001443
:114.19 g/mol
Boiling Pt:163…166 °C (760 mmHg)
Melting Pt:–38 °C
Density:0,93 g/cm³
Flash Pt:58 °C
Storage Temperature:Ambient
EINECS:209-512-0
UN:2617
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4610 to 1.4650 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together