Glycerine 86%, AnalaR NORMAPUR® analytical reagent
:56-81-5
:
:HOCH₂CH(OH)CH₂OH
:MFCD00004722
:92.09 g/mol
:13,04497
:Ambient
:200-289-5
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating)
- Assay:83.5 - 88.0%
- Acidity:Max. 0.0004 meq/g
- Alkalinity:Max. 0.0003 meq/g
- Density (20/4):1.220 - 1.230
- Esters of fatty acids (as C15H26O6):Max. 0.05 %
- Glyceraldehyde:Max. 30 ppm
- Heavy metals (as Pb):Max. 1 ppm
- Ignition residue (SO4):Max. 50 ppm
- Cl (Chloride):Max. 1 ppm
- NH4 (Ammonium):Max. 5 ppm
- SO4 (Sulphate):Max. 5 ppm
- Fe (Iron):Max. 1 ppm
- Water:13 - 16 %
Frequently Bought Together