Chromium ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 40842.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
Merck Index:12,02288
Boiling Pt:2469 °C (1013 hPa)
Melting Pt:1840 °C
Density:7,2 g/cm³ (20 °C)
:52 g/mol
Storage Temperature:Ambient
EINECS:231-157-5
CAS Number:7440-47-3
Formula:Cr
MDL Number:MFCD00010944