Zinc stabilised, powder, Supelco®
: Merck
:7440-66-6
:
:Zn
:MFCD00011291
:65.39 g/mol
:907 °C (1013 hPa)
:419,5 °C
:7,13 g/cm³ (20 °C)
:231-175-3
:9,III
:M-Clarity™ quality level = MQ200
:3077
:GHS09
:Ambient
- Solubility:(20 °C) (reaction),insoluble
- Melting Point:420 °C
- Molar Mass:65.39 g/mol
- Bulk Density:1800 - 2700 kg/m3
- Boiling Point:908 °C (1013 hPa)
- Vapor Pressure:1.33 hPa (487 °C)
- Density:7.14 g/cm3 (20 °C)
- pH:(H2O) not applicable
Frequently Bought Together