Zinc ≥99.9%, pellets, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7440-66-6
:
Formula:Zn
MDL Number:MFCD00011291
:65.39 g/mol
Merck Index:12,10255
Boiling Pt:908 °C (1013 hPa)
Melting Pt:420 °C
Density:7,14 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-175-3
Specification Test Results
- Assay:Min. 99.9 %
- Mean diameter:3 to 8 mm
- As (Arsenic):Max. 0.1 ppm
- Cd (Cadmium):Max. 30 ppm
- Cu (Copper):Max. 10 ppm
- Fe (Iron):Max. 0.003 %
- Pb (Lead):Max. 50 ppm
- Sn (Tin):Max. 50 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
spec_VWRC29080.238 spec_VWRC29080.295 spec_VWRC29080.410
Product specification
Frequently Bought Together