Order Entry
Northern Ireland
ContactUsLinkComponent
Zinc ≥99.8%, coarse powder, AnalaR NORMAPUR® ACS analytical reagent
Zinc ≥99.8%, coarse powder, AnalaR NORMAPUR® ACS analytical reagent
Catalog # 29068.236
Supplier:  VWR Chemicals
CAS Number:  
undefined
Zinc ≥99.8%, coarse powder, AnalaR NORMAPUR® ACS analytical reagent
Catalog # 29068.236
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay:
    Min. 99.8 %
  • As (Arsenic):
    Max. 0.1 ppm
  • Cd (Cadmium):
    Max. 20 ppm
  • Cu (Copper):
    Max. 10 ppm
  • Fe (Iron):
    Max. 100 ppm
  • Pb (Lead):
    Max. 50 ppm
  • Conforms to ACS:
    Passes test

Specification Test Results

About this item

 :
 :65.39 g/mol
Boiling Pt:907 °C (1013 hPa)
Melting Pt:419,5 °C
Density:7,13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-175-3
UN:1436
ADR:4.3 (4.2),II
 :GHS02
 :GHS09
CAS Number:7440-66-6
Formula:Zn
MDL Number:MFCD00011291