Zinc ≥99.9%, sticks, EMSURE® for analysis, Supelco®
1.08782.0500
: Merck
:
- Pack type:Plastic bag
- Pk:500 g
- Seq:3645
- Assay (complexometric):≥ 99.9 %
- As (Arsenic):≤ 0.00001 %
- Cd (Cadmium):≤ 0.005 %
- Cu (Copper):≤ 0.002 %
- Fe (Iron):≤ 0.005 %
- Pb (Lead):≤ 0.005 %
- Sn (Tin):≤ 0.001 %
:
:65.39 g/mol
:12,10255
:908 °C (1013 hPa)
:420 °C
:7,14 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:Ambient
:231-175-3
:7440-66-6
:Zn
:MFCD00011291