ContactUsLinkComponent
Sterilisation wraps, KIMGUARD* KC 200
Sterilisation wraps, KIMGUARD* KC 200
Catalog # KIMB68012
Supplier:  KIMBERLY CLARK
   Waste Reducing
Sterilisation wraps, KIMGUARD* KC 200
Catalog # KIMB68012
Supplier:  KIMBERLY CLARK
   Waste Reducing
Specifications
  • Colour:
    Light blue
  • Description:
    Regular-strength sterilisation wraps
  • Dimensions:
    300×300 mm
  • Seq:
    001
  • Environmentally Preferable:
    Waste Reducing
  • Pk:
    1.000
Specifications
About this item
Wraps with barrier and strength properties that help prevent contamination after sterilisation. These wraps are steam, EO and gas plasma compatible.
  • Made from low-linting SMS fabric
  • Sequential
  • Latex-free
Regular-strength wraps can be used to package linen towels, gowns and packs, light basin sets and single instruments.