Zinc ≥99.994% (metals basis), rods, Ø 3.18 mm (0.125 in)
Catalog # 43695.HO
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:350 mm
- Seq:1892
Specifications
About this item
:
:65.39 g/mol
Merck Index:12,10255
Boiling Pt:908 °C (1013 hPa)
Melting Pt:420 °C
Density:7,14 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-175-3
CAS Number:7440-66-6
Formula:Zn
MDL Number:MFCD00011291