Zinc ≥99.994% (metals basis), foil, Puratronic®, Thickness: 0.25 mm (0.01 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-66-6
:
Formula:Zn
MDL Number:MFCD00011291
:65.39 g/mol
Merck Index:12,10255
Boiling Pt:908 °C (1013 hPa)
Melting Pt:420 °C
Density:7,14 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-175-3
Frequently Bought Together