Zinc ≥99.9985% (metals basis), rods, Ø 7 mm (0.3 in)
Catalog # 11471.DM
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 mm
- Seq:1733
Specifications
About this item
:
:65.39 g/mol
Merck Index:12,10255
Boiling Pt:908 °C (1013 hPa)
Melting Pt:420 °C
Density:7,14 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-175-3
CAS Number:7440-66-6
Formula:Zn
MDL Number:MFCD00011291