Copper ≥99.999% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 40838.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:63.55 g/mol
Merck Index:13,02545
Boiling Pt:2595 °C (1013 hPa)
Melting Pt:1083 °C
Density:8,94 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-159-6
CAS Number:7440-50-8
Formula:Cu
MDL Number:MFCD00010965