
Barium ≥99.9% (metals basis), crystalline dendritic solid
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-39-3
:
Formula:Ba
MDL Number:MFCD00134031
:137.33 g/mol
Merck Index:12,00991
Boiling Pt:1640 °C (1013 hPa)
Melting Pt:725 °C
Density:3,51 g/cm³ (20 °C)
UN:1400
ADR:4.3,II
:ampouled under argon
EINECS:231-149-1
:GHS02
Storage Temperature:Ambient
Related Information
Frequently Bought Together