

Lithium, sticks in paraffin oil for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7439-93-2
:
Formula:Li
MDL Number:MFCD00134051
:6.94 g/mol
Merck Index:13,05542
Boiling Pt:1317 °C (1013 hPa)
Melting Pt:179 °C
Density:0,53 g/cm³ (20 °C)
UN:1415
ADR:4.3,I
:M-Clarity™ quality level = MQ200
EINECS:231-102-5
:GHS02
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:179 °C
- Molar Mass:6.94 g/mol
- Boiling Point:1347 °C
- Vapor Pressure:1 hPa (723 °C)
- Density:0.534 g/cm3 (20 °C)
- Hill Formula:Li
- Chemical Formula:Li
Specification Test Results
Frequently Bought Together