

Lithium 99% (metals basis), granules 1-6 mm (0.04-0.2 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-93-2
:
Formula:Li
MDL Number:MFCD00134051
:6.94 g/mol
Merck Index:13,05542
Boiling Pt:1317 °C (1013 hPa)
Melting Pt:179 °C
Density:0,53 g/cm³ (20 °C)
UN:1415
ADR:4.3,I
:Packaged under argon.
EINECS:231-102-5
:GHS02
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together