4,4'-Dimethylbenzophenone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:611-97-2
:
Formula:C₁₅H₁₄O
MDL Number:MFCD00017214
:210.28 g/mol
Boiling Pt:200 °C (17 mmHg)
Melting Pt:92…96 °C
Density:1,18 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-287-6
Related Information
Frequently Bought Together