ß-Nicotinamide adenine dinucleotide phosphate tetrasodium salt hydrate (NADPH-Na4 hydrate, reduced form) 96%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2646-71-1
:
Formula:C₂₁H₂₆N₇Na₄O₁₇P₃·xH₂O
MDL Number:MFCD10567216
:833.35 g/mol (anhy)
Storage Temperature:Freezer
EINECS:220-163-3
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Water:≤8 % (K.F.)
- HPLC:≥95 %
- Sodium (Na):10 to 12 %
Specification Test Results
Frequently Bought Together