Schiff reagent for analysis of aldehydes, Supelco®
Catalog # 1.09034.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay (SO₂):0.15 - 0.30 %
- Application test:conforms
- pH:1.5 (H2O, 20 °C)
Specification Test Results
About this item
:
Density:1,0 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
MDL Number:MFCD00081985