



Carbol fuchsin solution (Ziehl-Neelsen), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4197-24-4
:
Formula:C₂₁H₂₂ClN₃
MDL Number:MFCD18071399
:351.88 g/mol
Density:0,99 g/cm³ (20 °C)
Flash Pt:47 °C
Storage Temperature:Ambient
EINECS:211-189-6
UN:1992
ADR:3 (6.1),III
:M-Clarity™ quality level = MQ400
:GHS08
:GHS09
:GHS05
:GHS02
:1
Related Information
Certificates
Search by Lot NumberMERC1.09215.0100_TDS_EN MERC1.09215.0500_TDS_EN ProductPage_109215
Product specification
Merck The Color of Tissue Diagnostics
Product Literature
msds_109215
SDS
Frequently Bought Together