ContactUsLinkComponent
Carbol fuchsin, pure
Corrosive
Irritant
Health
Carbol fuchsin, pure
Certificates
undefined
Corrosive
Irritant
Health
Carbol fuchsin, pure
Specification Test Results
  • Appearance:
    Dark green Crystalline powder
  • Infrared spectrum:
    Conforms
  • UV spectrum:
    Complies
  • UV Lambda max:
    (in water) about 542 nm
  • Suitability:
    (for microscopy) Passes test