ContactUsLinkComponent
Lab tower, LT 828
Lab tower, LT 828
Catalog # 181-3110
Supplier:  KNF
undefined
Lab tower, LT 828
Catalog # 181-3110
Supplier:  KNF
Specifications
  • Flow rate:
    14 l/min
  • Description:
    Lab tower LT 828
  • Option:
    Aspirator
  • W×D×H:
    256×305×661 mm
  • Pk:
    1
Specifications
About this item
Multipurpose laboratory instrument for handling liquids. It can be used for filtering (in combination with a vacuum filtration unit), pipetting and aspirating liquids. The flow rate during aspirating and filtering, and the aspiration and dispensing speed when pipetting, can both be regulated from the control panel. A vacuum gauge indicates the pressure in the collecting bottle. The vacuum that the tower needs in order to work is created by a diaphragm vacuum pump. Liquid media are prevented from penetrating the pump by a protective sterile filter. The tower is equipped with locking wheels and fits under the laboratory workbench. It is equally convenient for left- and right-handed operation.
  • Very quiet running
  • Pipetting speed can be adjusted very precisely
  • Can be used with a range of pipettes and tips
  • Autoclavable, spill-proof glass bottle
  • Chemically resistant
Delivery: Delivery includes the tower with central housing, a pipetting device with sterile filter, an autoclavable collecting bottle, an aspirator, and a plug with connecting cable.