Digital stopwatch, bench, 5500
Desktop stopwatch, 1/100 s, 10 h, Black, 100×45×120 mm
Catalog # 609-0003
Marketsource
Supplier: EA COMBS
Jump to:
Product Details & Documents
Desktop stopwatch with bold LCD display.
- 1/100 second stopclock
- Lap and cumulative timing
- Counts up to 10 hours
- Three 4 mm sockets allow for external switching
: Supplied with AA battery.
Specifications
- Colour:Black
- Description:Desktop stopwatch, 1/100 s, 10 h
- W×D×H:100×45×120 mm



