Order Entry
Northern Ireland
ContactUsLinkComponent
 
Bottle-top dispensers, Pressmatic R
  613-4197
undefined
Bottle-top dispensers, Pressmatic R
  613-4197

 

  • Volume:
    10 - 60 ml
  • Accuracy:
    <0,1 %
  • Division:
    1,00 ml
  • Imprecision:
    \<0,7 %
  • Pk:
    1

 

 

Variable volume, autoclavable.

  • Ergonomic design with recirculation valve
  • Chemically resistant and non-sticking ceramic piston
  • Simple and accurate volume setting
  • Fully rotational on bottle for optimum positioning
  • Can be autoclaved at 121 °C

The Pressmatic R have an innovative design that maximises safety and ease of use. A recirculation valve means that reagent used for priming is delivered straight back into the bottle making it safer and less wasteful. Once the dispenser is fitted to the bottle, it is fully rotational for optimum positioning. The precise volume setting is by a self-locking rotating ring that is robust, easy to use and will not move during operation. The piston is manufactured from high-grade ceramic eliminating abrasion and sticking. The liquid pathway is made up of glass, PTFE and ceramic to give outstanding chemical resistance. The whole unit can be autoclaved without dismantling.

 : Supplied with three bottle adapters to fit most types of vessel and a works calibrated certificate.