Orders LinkContactUsLinkComponent
VWR®, Stirring Rods
VWR®, Stirring Rods
Supplier:   VWR Collection
VWR®, Stirring Rods
Supplier:  VWR Collection
Certificates
VWR®, Stirring Rods
VWR®, Stirring Rods
3 Options Available Below
Product Details & Documents
Solid PTFE.
  • Inert and will not scratch, can be used up to 280 °C
Product Family Options
Product Information
  • Material
    Ø
    Length
    Availability
    Price
  • PTFE
    8 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - £23.80
    £23.80
    1 * 5 items
       £23.80
  • PTFE
    8 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - £32.40
    £32.40
    1 * 5 items
       £32.40
  • PTFE
    8 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - £37.10
    £37.10
    1 * 5 items
       £37.10