Safety spectacles, uvex cybric
Catalog # 111-0146
Supplier: UVEX
Specifications
- Type:Anti-UV (2-1,2), anti-scratch, chemical-resistant, padded side arms
- Colour:Black/orange
- Seq:01
- Lenses:Clear PC
- Pk:1
Specifications
About this item
Sporty designed dual lens safety glasses with innovative lenses.
- Padded arms ensure a pressure-free and comfortable fit
- Wide field of vision
Weight: 48 g
EN 166, EN 168, EN 170
Certifications: EN 166, EN 168, EN 170