ContactUsLinkComponent
Dispensers for Parafilm® M sealing film, Nalgene®
Dispensers for Parafilm® M sealing film, Nalgene®
Catalog # 291-0013
   Low Manufacturing Impact
undefined
Dispensers for Parafilm® M sealing film, Nalgene®
Catalog # 291-0013
   Low Manufacturing Impact
Specifications
  • Description:
    PARAFILM® M dispenser
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1
Specifications
About this item
For clean storage and easy dispensing of Parafilm® M sealing film.
  • Designed for two rolls 50 mm wide or one roll 100 mm wide
  • Easy to clean
  • Clear acrylic