Test tube racks, radioimmunassay, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
PP, white.
- Temperature resistant to 121 °C, steam sterilisable
- Good chemical resistance
These racks provide a practical method of pipetting and storing a large number of samples. By using alternate rows and rotating the tubes, it is easy to keep track of the last tube filled. Also useful in clinical laboratories for storage of samples.
Frequently Bought Together