ContactUsLinkComponent
Gas dispersion tube
Gas dispersion tube
Catalog # 511-0818
Supplier:  DIXON SCIENTIFIC
undefined
Gas dispersion tube
Catalog # 511-0818
Supplier:  DIXON SCIENTIFIC
Specifications
  • Head Ø:
    10 mm
  • Porosity:
    P160
  • Length:
    120 mm
  • Pk:
    1
Specifications
About this item
This gas dispersion tube offers relatively high resistance to temperature changes.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion