Flasks, round bottom with two necks and Quickfit® ground joints
Supplier: SciLabware
Certificates
About this item
PYREX®, borosilicate glass, with two necks, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
One vertical centre neck and one angled side neck. With matt white labelling area.
Certifications: ISO 1773, BS 6352
Frequently Bought Together