Erlenmeyer flasks, narrow neck with strengthened rim
Supplier: DIXON SCIENTIFIC
Certificates
About this item
Borosilicate glass, grade 3.3.
- Very good chemical resistance
- High temperature resistance
- Reduced breakage
For a wide range of laboratory applications. Volume graduations and labelling area.
Frequently Bought Together