Orders LinkContactUsLinkComponent
Groundwater sampling devices, AquaPrep™
Groundwater sampling devices, AquaPrep™
 :  
Groundwater sampling devices, AquaPrep™
Groundwater sampling devices, AquaPrep™
AquaPrep™ devices are designed for the preparation of groundwater samples, with relatively low levels of particulate matter, for dissolved metals analysis and to provide a cost effective alternative to the GWV high capacity capsules for light sediments.
  • Meets filtration requirements of the U.S. EPA. for 0,45 µm filtration
  • Self-contained, disposable units eliminate the time and hazards associated with cleaning filter holders
AquaPrep device: Thermopor membrane (polyester reinforced PS), PP housing.
AquaPrep™-V device: Versapor® membrane (acrylic copolymer on a non woven support), PP housing.