Bottle top filters, VacuCap®
Supplier: Cytiva (Formerly Pall Lab)
Certificates
About this item
Supor® hydrophilic polyethersulphone (PES) membrane; PP housing. VacuCap® filtration device eliminates the need to constantly refill funnel in traditional bottle top filters as it draws directly from the mixing reservoir. The patented small design accepts a variety of collection vessels and reduces laboratory storage space and waste. The Supor® membrane in the VacuCap® 60 and VacuCap® 90 devices provides high flow rates for fast vacuum filtration of 100 ml to 5 litres of aqueous solutions and is available with prefilter to increase throughput of particulate laden solutions.
- Excellent for cell culture media preparation and available with 0,1 µm pore size to remove mycoplasma
- Pre-sterilised by gamma irradiation to eliminate contamination by EtO residuals, individually packed
- Environmentally friendly design with minimal plastic waste
Specifications
- Catalog No:
- 515-0014
- 515-0015
- 515-0019
- 515-0017
- 516-9813
- 515-0016
- 515-0018
- 516-9812
- 515-0012
- 515-0013
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
Specifications
Related Information
Certificates
Search by Lot NumberPall Laboratory VacuCap® Vacuum Filtration Devices
Product Literature
Frequently Bought Together